Ethyl-2-methoxyisonicotinate 97%

BLDPBD236512-5GEA 142 EUR
BLDPBD236512-5G BLDPBD236512-25G BLDPBD236512-1G
Ethyl-2-methoxyisonicotinate 97%
Ethyl-2-methoxyisonicotinate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₁NO₃
MW: 181,1900 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11504865
Núm. CAS: 105596-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR