3-(Hydroxymethyl)phenylboronic acid pinacol ester 97%
|
|
Sinónimos:
3-(Hydroxymethyl)benzeneboronic acid pinacol ester
BLDPBD236258-250MGEA
6
EUR
BLDPBD236258-250MG
BLDPBD236258-10G
BLDPBD236258-100G
BLDPBD236258-5G
BLDPBD236258-25G
BLDPBD236258-1G
3-(Hydroxymethyl)phenylboronic acid pinacol ester 97%
3-(Hydroxymethyl)phenylboronic acid pinacol ester
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₃H₁₉BO₃ MW: 234,10 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09266196 Núm. CAS: 443776-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...