3,5-Dimethylcyclohexanol (mixture of isomers) 98%
BLDPBD235175-25GEA 119 EUR
BLDPBD235175-25G BLDPBD235175-100G BLDPBD235175-1G BLDPBD235175-5G
3,5-Dimethylcyclohexanol (mixture of isomers) 98%
3,5-Dimethylcyclohexanol (mixture of isomers)
Storage condition: Sealed in dry, Room Temperature
Formula: C₈H₁₆O
MW: 128,2120 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001447
Núm. CAS: 5441-52-1