4-Bromo-1-chloro-2-ethylbenzene 95+%

BLDPBD232084-5GEA 79 EUR
BLDPBD232084-5G BLDPBD232084-1G
4-Bromo-1-chloro-2-ethylbenzene 95+%
4-Bromo-1-chloro-2-ethylbenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈BrCl
MW: 219,51 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00672954
Núm. CAS: 289039-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR