4-Bromo-N,N-dimethyl-2-fluorobenzamide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD231567-5GEA 200 EUR
BLDPBD231567-5G BLDPBD231567-250MG BLDPBD231567-1G BLDPBD231567-10G
4-Bromo-N,N-dimethyl-2-fluorobenzamide 98%
4-Bromo-N,N-dimethyl-2-fluorobenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉BrFNO
MW: 246,0763 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09878360
Núm. CAS: 749927-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR