4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD231489-250MGEA 90 EUR
BLDPBD231489-250MG BLDPBD231489-1G
4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid 98%
4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₄H₂₀BNO₃
MW: 261,1300 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09027224
Núm. CAS: 913835-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR