4-Amino-2,5-dichloropyridine 98%

BLDPBD231155-1GEA 85 EUR
BLDPBD231155-1G BLDPBD231155-5G BLDPBD231155-250MG BLDPBD231155-100MG
4-Amino-2,5-dichloropyridine 98%
4-Amino-2,5-dichloropyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₄Cl₂N₂
MW: 163,005 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD11044256
Núm. CAS: 405230-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR