3-Bromo-2,6-difluorophenol 98%

BLDPBD231023-250MGEA 42 EUR
BLDPBD231023-250MG BLDPBD231023-1G BLDPBD231023-5G
3-Bromo-2,6-difluorophenol 98%
3-Bromo-2,6-difluorophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₃BrF₂O
MW: 208,9882 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD12910487
Núm. CAS: 221220-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR