6-Bromo-7-methylimidazo[1,2-a]pyridine 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD230263-1GEA 51 EUR
BLDPBD230263-1G BLDPBD230263-25G BLDPBD230263-250MG BLDPBD230263-5G
6-Bromo-7-methylimidazo[1,2-a]pyridine 95%
6-Bromo-7-methylimidazo[1,2-a]pyridine

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₇BrN₂
MW: 211,0600 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09258769
Núm. CAS: 116355-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR