5-Bromo-4-methyl-2-nitroaniline 98%

BLDPBD229625-1GEA 61 EUR
BLDPBD229625-1G BLDPBD229625-250MG BLDPBD229625-5G BLDPBD229625-25G
5-Bromo-4-methyl-2-nitroaniline 98%
5-Bromo-4-methyl-2-nitroaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₇BrN₂O₂
MW: 231,0500 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00156314
Núm. CAS: 40371-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR