4-(N,N-Diphenylamino)phenylboronic acid pinacol ester 97%

BLDPBD228350-5GEA 61 EUR
BLDPBD228350-5G BLDPBD228350-25G BLDPBD228350-1G BLDPBD228350-100G BLDPBD228350-250MG
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester 97%
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₄H₂₆BNO₂
MW: 371,29 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD13195770
Núm. CAS: 267221-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR