2-Amino-1-naphthalenesulphonic acid 98%
|
|
BLDPBD22705-500GEA
23
EUR
BLDPBD22705-500G
BLDPBD22705-25G
BLDPBD22705-100G
2-Amino-1-naphthalenesulphonic acid 98%
2-Amino-1-naphthalenesulphonic acid
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₁₀H₉NO₃S MW: 223,25 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00003988 Núm. CAS: 81-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...