4-Bromo-3-methyl-1H-pyrazole 97%
|
|
BLDPBD2262-100GEA
118
EUR
BLDPBD2262-100G
BLDPBD2262-1G
BLDPBD2262-10G
BLDPBD2262-500G
BLDPBD2262-5G
BLDPBD2262-25G
4-Bromo-3-methyl-1H-pyrazole 97%
4-Bromo-3-methyl-1H-pyrazole
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₄H₅BrN₂ MW: 161,01 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00005241 Núm. CAS: 13808-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...