3-Boronobenzenesulphonamide pinacol ester 95%
|
|
BLDPBD224528-100MGEA
31
EUR
BLDPBD224528-100MG
BLDPBD224528-5G
BLDPBD224528-250MG
BLDPBD224528-1G
3-Boronobenzenesulphonamide pinacol ester 95%
3-Boronobenzenesulphonamide pinacol ester
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₂H₁₈BNO₄S MW: 283,1516 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07781217 Núm. CAS: 486422-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...