3-Chloro-5-methylphenylboronic acid 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD222894-1GEA 45 EUR
BLDPBD222894-1G BLDPBD222894-100MG BLDPBD222894-5G BLDPBD222894-250MG
3-Chloro-5-methylphenylboronic acid 97%
3-Chloro-5-methylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₈BClO₂
MW: 170,40 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09027241
Núm. CAS: 913836-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR