6-Bromo-4-methylquinoline 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD222159-100MGEA 15 EUR
BLDPBD222159-100MG BLDPBD222159-5G BLDPBD222159-25G BLDPBD222159-10G BLDPBD222159-250MG BLDPBD222159-1G
6-Bromo-4-methylquinoline 97%
6-Bromo-4-methylquinoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₈BrN
MW: 222,08 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09258893
Núm. CAS: 41037-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR