2-Ethoxybenzonitrile 95%

BLDPBD22072-1GEA 6 EUR
BLDPBD22072-1G BLDPBD22072-10G BLDPBD22072-25G BLDPBD22072-5G BLDPBD22072-100G
2-Ethoxybenzonitrile 95%
2-Ethoxybenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉NO
MW: 147,18 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001789
Núm. CAS: 6609-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR