2-Amino-6-bromoquinoline 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD219355-5GEA 389 EUR
BLDPBD219355-5G BLDPBD219355-1G BLDPBD219355-250MG
2-Amino-6-bromoquinoline 98%
2-Amino-6-bromoquinoline
Formula: C₉H₇BrN₂
MW: 223,0700 g/mol
Número MDL: MFCD06738670
Núm. CAS: 791626-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR