3-(Dimethylamino)-1,2-propanediol 98%

BLDPBD21852-10GEA 7 EUR
BLDPBD21852-10G BLDPBD21852-0.025G BLDPBD21852-5G BLDPBD21852-25G
3-(Dimethylamino)-1,2-propanediol 98%
3-(Dimethylamino)-1,2-propanediol

Storage condition: Sealed in dry, Room Temperature

Formula: C5H13NO2
MW: 119,1622 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00004714
Núm. CAS: 623-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR