Ácido 5-formilsalicílico 98% (by HPLC)

BLDPBD21803-100GEA 117 EUR
BLDPBD21803-100G BLDPBD21803-500G BLDPBD21803-25G BLDPBD21803-5G BLDPBD21803-10G BLDPBD21803-1000G
Ácido 5-formilsalicílico 98% (by HPLC)
Ácido 5-formilsalicílico

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₆O₄
MW: 166,1308 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006945
Núm. CAS: 616-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR