2,6-Dichloro-4-methylaniline 98%

BLDPBD217791-10GEA 499 EUR
BLDPBD217791-10G BLDPBD217791-1G BLDPBD217791-5G BLDPBD217791-250MG
2,6-Dichloro-4-methylaniline 98%
2,6-Dichloro-4-methylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₇Cl₂N
MW: 176,043 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00190134
Núm. CAS: 56461-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR