5-Bromo-2-chloroaniline 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD21737-500GEA 487 EUR
BLDPBD21737-500G BLDPBD21737-1G BLDPBD21737-10G BLDPBD21737-5G BLDPBD21737-100G BLDPBD21737-25G
5-Bromo-2-chloroaniline 98%
5-Bromo-2-chloroaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₅BrClN
MW: 206,47 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06796283
Núm. CAS: 60811-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR