(R)-(-)-Glycidyl butyrate 95%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD21720-5GEA
7
EUR
BLDPBD21720-5G
BLDPBD21720-25G
BLDPBD21720-500G
BLDPBD21720-100G
(R)-(-)-Glycidyl butyrate 95%
(R)-(-)-Glycidyl butyrate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₇H₁₂O₃ MW: 144,17 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00075120 Núm. CAS: 60456-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...