3-Bromo-4-iodoanísol 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD216954-100GEA 969 EUR
BLDPBD216954-100G BLDPBD216954-5G BLDPBD216954-10G BLDPBD216954-1G BLDPBD216954-250MG BLDPBD216954-25G
3-Bromo-4-iodoanísol 97%
3-Bromo-4-iodoanísol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₆BrIO
MW: 312,9304 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD12547848
Núm. CAS: 466639-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR