Pyridazine-4,5-dicarboxylic acid 98%
|
|
BLDPBD21677-25GEA
682
EUR
BLDPBD21677-25G
BLDPBD21677-5G
BLDPBD21677-10G
BLDPBD21677-1G
Pyridazine-4,5-dicarboxylic acid 98%
Pyridazine-4,5-dicarboxylic acid
Storage condition: Sealed in dry, 2-8°C
Formula:
C₆H₄N₂O₄ MW: 168,1070 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD06742784 Núm. CAS: 59648-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...