3-Chloro-1H-pyrazole 98%

BLDPBD215704-250MGEA 10 EUR
BLDPBD215704-250MG BLDPBD215704-25G BLDPBD215704-5G BLDPBD215704-100MG BLDPBD215704-1G BLDPBD215704-10G
3-Chloro-1H-pyrazole 98%
3-Chloro-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formula: C₃H₃ClN₂
MW: 102,522 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD12756597
Núm. CAS: 14339-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR