2-Fluoro-4-formylphenylboronic acid 98%
|
|
BLDPBD215693-1GEA
18
EUR
BLDPBD215693-1G
BLDPBD215693-100G
BLDPBD215693-25G
BLDPBD215693-5G
BLDPBD215693-250MG
2-Fluoro-4-formylphenylboronic acid 98%
2-Fluoro-4-formylphenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₆BFO₃ MW: 167,9300 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07367058 Núm. CAS: 871126-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...