Methyl-3-bromo-5-(trifluoromethyl)benzoate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD215285-10GEA 80 EUR
BLDPBD215285-10G BLDPBD215285-100G BLDPBD215285-25G BLDPBD215285-1G BLDPBD215285-5G
Methyl-3-bromo-5-(trifluoromethyl)benzoate 98%
Methyl-3-bromo-5-(trifluoromethyl)benzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₆BrF₃O₂
MW: 283,0420 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11847338
Núm. CAS: 187331-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR