α,4-Dibromo-3-fluorotoluene 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD215160-25GEA 90 EUR
BLDPBD215160-25G BLDPBD215160-5G BLDPBD215160-1G BLDPBD215160-250MG BLDPBD215160-10G
α,4-Dibromo-3-fluorotoluene 97%
α,4-Dibromo-3-fluorotoluene

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₅Br₂F
MW: 267,92 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11847179
Núm. CAS: 127425-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR