4-Chloro-6-methyl-2-(methylthio)pyrimidine 98%

BLDPBD215159-250MGEA 8 EUR
BLDPBD215159-250MG BLDPBD215159-10G BLDPBD215159-100G BLDPBD215159-1G BLDPBD215159-25G BLDPBD215159-5G
4-Chloro-6-methyl-2-(methylthio)pyrimidine 98%
4-Chloro-6-methyl-2-(methylthio)pyrimidine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₇ClN₂S
MW: 174,65 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00023233
Núm. CAS: 17119-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR