Methyl 2-iodo-6-methylbenzoate 90%

BLDPBD214214-1GEA 102 EUR
BLDPBD214214-1G BLDPBD214214-250MG BLDPBD214214-5G
Methyl 2-iodo-6-methylbenzoate 90%
Methyl 2-iodo-6-methylbenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₉IO₂
MW: 276,071 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08061917
Núm. CAS: 103440-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR