3'-Bromo-2,2,2-trifluoroacetophenone 99%
|
|
BLDPBD213208-5GEA
121
EUR
BLDPBD213208-5G
BLDPBD213208-100MG
BLDPBD213208-25G
BLDPBD213208-1G
BLDPBD213208-250MG
BLDPBD213208-100G
3'-Bromo-2,2,2-trifluoroacetophenone 99%
3'-Bromo-2,2,2-trifluoroacetophenone
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₄BrF₃O MW: 253,0160 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00511232 Núm. CAS: 655-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...