1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
|
|
BLDPBD208755-25GEA
1125
EUR
BLDPBD208755-25G
BLDPBD208755-250MG
BLDPBD208755-1G
BLDPBD208755-10G
BLDPBD208755-5G
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₃H₂₁BN₂O₄ MW: 280,1278 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD08706026 Núm. CAS: 864754-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...