6-Bromonicotinamide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD208431-1GEA 40 EUR
BLDPBD208431-1G BLDPBD208431-5G BLDPBD208431-250MG
6-Bromonicotinamide 98%
6-Bromonicotinamide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₅BrN₂O
MW: 201,0200 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00266840
Núm. CAS: 889676-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR