4,7,10-Trioxa-1,13-tridecanediamine 95%

BLDPBD20792-100GEA 43 EUR
BLDPBD20792-100G BLDPBD20792-5G BLDPBD20792-25G BLDPBD20792-10G
4,7,10-Trioxa-1,13-tridecanediamine 95%
4,7,10-Trioxa-1,13-tridecanediamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₂₄N₂O₃
MW: 220,3092 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00059850
Núm. CAS: 4246-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR