(Methylthio)acetaldehyde dimethyl acetal 98%
|
|
BLDPBD20654-5GEA
26
EUR
BLDPBD20654-5G
BLDPBD20654-25G
(Methylthio)acetaldehyde dimethyl acetal 98%
(Methylthio)acetaldehyde dimethyl acetal
Storage condition: Sealed in dry, Room Temperature
Formula:
C₅H₁₂O₂S MW: 136,21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00009848 Núm. CAS: 40015-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...