2-Bromo-9,9-dimethylfluorene 98%

BLDPBD19952-5GEA 7 EUR
BLDPBD19952-5G BLDPBD19952-25G BLDPBD19952-1G BLDPBD19952-500G BLDPBD19952-100G
2-Bromo-9,9-dimethylfluorene 98%
2-Bromo-9,9-dimethylfluorene

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₅H₁₃Br
MW: 273,17 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09261272
Núm. CAS: 28320-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR