Ethyl thiophene-2-carboxylate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD19937-25GEA 30 EUR
BLDPBD19937-25G BLDPBD19937-500G BLDPBD19937-5G BLDPBD19937-100G
Ethyl thiophene-2-carboxylate 98%
Ethyl thiophene-2-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₈O₂S
MW: 156,20 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00005436
Núm. CAS: 2810-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR