N'-(Etilcarbonoimidoil)-N,N-dimetilpropano-1,3-diamina monoclorhidrato 99%

Sinónimos: EDC HClEDC

BLDPBD19757-5GEA 6 EUR
BLDPBD19757-5G BLDPBD19757-25G BLDPBD19757-10G
N'-(Etilcarbonoimidoil)-N,N-dimetilpropano-1,3-diamina monoclorhidrato 99%
N'-(Etilcarbonoimidoil)-N,N-dimetilpropano-1,3-diamina monoclorhidrato

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formula: C₈H₁₇N₃·HCl
MW: 191,7 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00012503
Núm. CAS: 25952-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR