6-Bromo-2-chloro-3-methylbenzeneboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD196763-5GEA 113 EUR
BLDPBD196763-5G BLDPBD196763-250MG BLDPBD196763-1G
6-Bromo-2-chloro-3-methylbenzeneboronic acid 98%
6-Bromo-2-chloro-3-methylbenzeneboronic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₇BBrClO₂
MW: 249,297 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09475910
Núm. CAS: 957120-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR