Methyl-4-amino-3-fluorobenzoate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD196503-25GEA 49 EUR
BLDPBD196503-25G BLDPBD196503-5G BLDPBD196503-10G BLDPBD196503-100G BLDPBD196503-1G
Methyl-4-amino-3-fluorobenzoate 98%
Methyl-4-amino-3-fluorobenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₈FNO₂
MW: 169,1530 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08689703
Núm. CAS: 185629-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR