Methyl-6-bromo-1H-indazole-4-carboxylate 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD196225-100GEA 767 EUR
BLDPBD196225-100G BLDPBD196225-5G BLDPBD196225-25G BLDPBD196225-250MG BLDPBD196225-1G
Methyl-6-bromo-1H-indazole-4-carboxylate 97%
Methyl-6-bromo-1H-indazole-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₇BrN₂O₂
MW: 255,0700 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07781319
Núm. CAS: 885518-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR