4-Bromo-1H-pyrazole 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD19300-10GEA 6 EUR
BLDPBD19300-10G BLDPBD19300-100G BLDPBD19300-25G BLDPBD19300-500G BLDPBD19300-1000G
4-Bromo-1H-pyrazole 98%
4-Bromo-1H-pyrazole

Storage condition: Inert atmosphere, Room Temperature

Formula: C₃H₃BrN₂
MW: 146,97 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00075602
Núm. CAS: 2075-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR