DL-Glutamic acid monohydrate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD19107-1000GEA 177 EUR
BLDPBD19107-1000G BLDPBD19107-500G BLDPBD19107-5G BLDPBD19107-100G BLDPBD19107-25G
DL-Glutamic acid monohydrate 98%
DL-Glutamic acid monohydrate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₉NO₄·1H₂O
MW: 165,15 g/mol (147,13 anhy)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00150703
Núm. CAS: 19285-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR