Hesperidin 97%
|
|
BLDPBD1901-5GEA
8
EUR
BLDPBD1901-5G
BLDPBD1901-500G
BLDPBD1901-10G
BLDPBD1901-100G
BLDPBD1901-25G
BLDPBD1901-1G
Hesperidin 97%
Hesperidin
Storage condition: Sealed in dry, 2-8°C
Formula:
C₂₈H₃₄O₁₅ MW: 610,56 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00075663 Núm. CAS: 520-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...