6-Hydroxy-7-methoxyquinazolin-4(1H)-one 98%

BLDPBD18955-25GEA 47 EUR
BLDPBD18955-25G BLDPBD18955-5G BLDPBD18955-250MG BLDPBD18955-10G BLDPBD18955-100G BLDPBD18955-500G BLDPBD18955-1G
6-Hydroxy-7-methoxyquinazolin-4(1H)-one 98%
6-Hydroxy-7-methoxyquinazolin-4(1H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈N₂O₃
MW: 192,1714 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07787563
Núm. CAS: 179688-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR