N-Carbobenzoxy-D-2-phenylglycine 98%

BLDPBD18908-1GEA 7 EUR
BLDPBD18908-1G BLDPBD18908-5G BLDPBD18908-100G BLDPBD18908-25G
N-Carbobenzoxy-D-2-phenylglycine 98%
N-Carbobenzoxy-D-2-phenylglycine

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₆H₁₅NO₄
MW: 285,30 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064977
Núm. CAS: 17609-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR