3-Pyridyloxyacetic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD183882-1GEA 190 EUR
BLDPBD183882-1G BLDPBD183882-5G BLDPBD183882-100MG BLDPBD183882-250MG
3-Pyridyloxyacetic acid 98%
3-Pyridyloxyacetic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₇NO₃
MW: 153,14 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04039772
Núm. CAS: 86649-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR