4'-Hydroxyheptanophenone 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD18351-100GEA 29 EUR
BLDPBD18351-100G BLDPBD18351-25G BLDPBD18351-1000G BLDPBD18351-500G BLDPBD18351-0.025G BLDPBD18351-10G
4'-Hydroxyheptanophenone 98%
4'-Hydroxyheptanophenone

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₁₈O₂
MW: 206,2800 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00191523
Núm. CAS: 14392-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR