2,4,5-Trifluoroiodobenzene 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD183333-25GEA 76 EUR
BLDPBD183333-25G BLDPBD183333-100G BLDPBD183333-10G BLDPBD183333-5G BLDPBD183333-1G
2,4,5-Trifluoroiodobenzene 97%
2,4,5-Trifluoroiodobenzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₂F₃I
MW: 257,98 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD04973898
Núm. CAS: 17533-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR